Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LECT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_531913.6 | XP_531913.1 | 2 - 3 | 359 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |