Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022421156.1 | XP_022276864.1 | 4 - 5 | 1918 | 90 | |
XM_537221.6 | XP_537221.1 | 3 - 4 | 2024 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |