Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621115.3 | XP_005621172.1 | 594 | 75 | ||
XM_022419726.1 | XP_022275434.1 | 574 | 75 | |||
XM_022419727.1 | XP_022275435.1 | 565 | 75 | |||
XM_536880.6 | XP_536880.2 | 714 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris |