Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195403.1 | NP_001182332.1 | 3 - 4 | 462 | 126 | |
NM_001195825.1 | NP_001182754.1 | 3 - 4 | 471 | 126 | ||
NM_001195826.1 | NP_001182755.1 | 3 - 4 | 421 | 126 | ||
NM_001195827.1 | NP_001182756.1 | 3 - 4 | 412 | 126 | ||
XM_005631053.3 | XP_005631110.1 | 4 - 5 | 560 | 126 | ||
XM_014120867.1 | XP_013976342.1 | 3 - 4 | 931 | 126 | ||
XM_014120868.2 | XP_013976343.1 | 4 - 5 | 435 | 126 | ||
XM_022405072.1 | XP_022260780.1 | 4 - 5 | 569 | 126 | ||
XM_022405074.1 | XP_022260782.1 | 3 - 4 | 423 | 126 | ||
XM_022405075.1 | XP_022260783.1 | 3 - 4 | 405 | 126 | ||
GenBank mRNA | FJ159123.1 | - | 3 - 4 | 390 | 126 | |
FJ159124.1 | - | 3 - 4 | 408 | 126 | ||
FJ159125.1 | - | 3 - 4 | 399 | 126 | ||
FJ159126.1 | - | 3 - 4 | 417 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |