Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426844.1 | XP_022282552.1 | 7 - 8 | 2172 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |