Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426279.1 | XP_022281987.1 | 4 - 5 | 520 | 118 | |
XM_539015.6 | XP_539015.1 | 4 - 5 | 680 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |