Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_538517.6 | XP_538517.2 | 5 - 6 | 717 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |