Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRODH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313832.1 | NP_001300761.1 | 7 - 8 | 1097 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |