Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638422.3 | XP_005638479.1 | 1 - 2 | 164 | 87 | |
XM_022412585.1 | XP_022268293.1 | 2 - 3 | 158 | 87 | ||
XM_022412586.1 | XP_022268294.1 | 2 - 3 | 147 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |