Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_844610.5 | XP_849703.1 | 1 - 2 | 251 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |