Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621395.3 | XP_005621452.1 | 3 - 4 | 539 | 98 | |
XM_845033.5 | XP_850126.1 | 3 - 4 | 590 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |