Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022406176.1 | XP_022261884.1 | 3 - 4 | 368 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |