Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005622754.3 | XP_005622811.1 | 3 - 4 | 480 | 92 | |
XM_005622755.3 | XP_005622812.1 | 3 - 4 | 763 | 92 | ||
XM_005622756.3 | XP_005622813.1 | 3 - 4 | 635 | 92 | ||
XM_014115357.2 | XP_013970832.1 | 3 - 4 | 623 | 92 | ||
XM_846978.5 | XP_852071.1 | 3 - 4 | 571 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |