Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_845219.5 | XP_850312.1 | 846 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris |