Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022423640.1 | XP_022279348.1 | 8 - 9 | 811 | 123 | |
XM_022423641.1 | XP_022279349.1 | 8 - 9 | 821 | 123 | ||
XM_845214.3 | XP_850307.1 | 8 - 9 | 789 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |