Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104959.1 | NP_001098429.1 | 1 - 2 | 30 | 85 | |
XM_005631766.2 | XP_005631823.1 | 1 - 2 | 577 | 85 | ||
GenBank mRNA | AB299409.1 | - | 1 - 2 | 30 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |