Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022405407.1 | XP_022261115.1 | 2 - 3 | 699 | 67 | |
XM_847563.5 | XP_852656.2 | 2 - 3 | 692 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |