Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005635177.3 | XP_005635234.1 | 1 - 2 | 186 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Canis familiaris Probe spans exons |