Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACRV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619589.3 | XP_005619646.1 | 158 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |