Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639430.3 | XP_005639487.1 | 1 | 143 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |