Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631904.3 | XP_005631961.1 | 3 - 4 | 399 | 85 | |
XM_541001.5 | XP_541001.2 | 3 - 4 | 520 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |