Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_535665.6 | XP_535665.2 | 1 - 2 | 148 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |