Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_845166.5 | XP_850259.1 | 2 - 3 | 1352 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |