Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616116.3 | XP_005616173.1 | 1 - 2 | 155 | 64 | |
XM_845668.5 | XP_850761.1 | 1 - 2 | 155 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |