Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014118906.2 | XP_013974381.1 | 2 | 326 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |