Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629298.3 | XP_005629355.1 | 12 - 13 | 1373 | 83 | |
XM_014119382.2 | XP_013974857.1 | 17 - 18 | 2384 | 83 | ||
XM_022403436.1 | XP_022259144.1 | 14 - 15 | 1606 | 83 | ||
XM_022403437.1 | XP_022259145.1 | 13 - 14 | 1404 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |