Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623216.3 | XP_005623273.1 | 2 | 4429 | 145 | |
XM_005623217.3 | XP_005623274.1 | 2 | 4429 | 145 | ||
XM_014115750.2 | XP_013971225.1 | 2 | 4429 | 145 | ||
XM_547725.5 | XP_547725.2 | 2 | 4429 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |