Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_534436.6 | XP_534436.4 | 5 - 6 | 716 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons |