Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426029.1 | XP_022281737.1 | 3 | 825 | 200 | |
XM_022426030.1 | XP_022281738.1 | 3 | 828 | 200 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |