Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_548071.6 | XP_548071.2 | 1488 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |