Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197169.1 | NP_001184098.1 | 2 | 140 | 110 | |
XM_005615765.3 | XP_005615822.1 | 3 | 300 | 110 | ||
GenBank mRNA | HM135452.1 | - | 2 | 140 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |