Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_844418.5 | XP_849511.1 | 3 - 4 | 874 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |