Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005622773.1 | XP_005622830.1 | 4 | 1497 | 66 | |
XM_005622774.1 | XP_005622831.1 | 4 | 1480 | 66 | ||
XM_022421321.1 | XP_022277029.1 | 3 | 1476 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |