Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014122455.2 | XP_013977930.1 | 760 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris |