Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003433569.4 | XP_003433617.1 | 2 - 3 | 374 | 107 | |
XM_005637097.3 | XP_005637154.1 | 2 - 3 | 379 | 107 | ||
XM_005637098.3 | XP_005637155.1 | 3 - 4 | 578 | 107 | ||
XM_005637099.3 | XP_005637156.1 | 3 - 4 | 677 | 107 | ||
XM_005637100.3 | XP_005637157.1 | 3 - 4 | 644 | 107 | ||
XM_005637101.3 | XP_005637158.1 | 2 - 3 | 408 | 107 | ||
XM_005637102.3 | XP_005637159.1 | 2 - 3 | 515 | 107 | ||
XM_005637103.3 | XP_005637160.1 | 2 - 3 | 658 | 107 | ||
XM_014108640.2 | XP_013964115.1 | 2 - 3 | 882 | 107 | ||
XM_022411066.1 | XP_022266774.1 | 3 - 4 | 475 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons |