Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618393.3 | XP_005618450.3 | 2 - 3 | 415 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons |