Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417270.1 | XP_022272978.1 | 3 | 705 | 199 | |
XM_545973.6 | XP_545973.3 | 2 | 734 | 199 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |