Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623590.3 | XP_005623647.1 | 2 | 1136 | 172 | |
XM_005623591.3 | XP_005623648.1 | 2 | 1147 | 172 | ||
XM_022422180.1 | XP_022277888.1 | 2 | 1140 | 172 | ||
XM_022422181.1 | XP_022277889.1 | 2 | 1026 | 172 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |