Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGFG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107538.2 | XP_013963013.1 | 4 - 5 | 390 | 174 | |
XM_022410057.1 | XP_022265765.1 | 4 - 5 | 390 | 174 | ||
XM_022410058.1 | XP_022265766.1 | 4 - 5 | 513 | 174 | ||
XM_022410059.1 | XP_022265767.1 | 4 - 5 | 390 | 174 | ||
XM_022410060.1 | XP_022265768.1 | 4 - 5 | 390 | 174 | ||
XM_022410061.1 | XP_022265769.1 | 4 - 5 | 390 | 174 | ||
XM_022410062.1 | XP_022265770.1 | 4 - 5 | 390 | 174 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons |