Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_542135.4 | XP_542135.2 | 5 | 1124 | 175 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |