Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287105.1 | NP_001274034.1 | 2 | 230 | 141 | |
NM_001287106.1 | NP_001274035.1 | 2 | 201 | 141 | ||
GenBank mRNA | CO658560.1 | - | 4 | 380 | 141 | |
DN863506.1 | - | 4 | 395 | 141 | ||
DN871196.1 | - | 2 | 201 | 141 | ||
GR889127.1 | - | 2 | 231 | 141 | ||
GR894797.1 | - | 2 | 179 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Amplicon spans exons and probe does not span exons |