Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619427.3 | XP_005619484.1 | 4 | 1043 | 96 | |
XM_022417965.1 | XP_022273673.1 | 4 | 1002 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |