Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638005.3 | XP_005638062.1 | 5 - 6 | 1102 | 112 | |
XM_544093.6 | XP_544093.3 | 6 - 7 | 1168 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |