Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617139.3 | XP_005617196.1 | 383 | 143 | ||
XM_014122653.2 | XP_013978128.1 | 396 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris |