Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640948.1 | XP_005641005.1 | 2 | 272 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |