Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022406737.1 | XP_022262445.1 | 1 | 539 | 89 | |
XM_845960.5 | XP_851053.1 | 1 | 539 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |