Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014120496.2 | XP_013975971.1 | 4 - 5 | 407 | 86 | |
XM_014120497.2 | XP_013975972.1 | 4 - 5 | 389 | 86 | ||
XM_022404741.1 | XP_022260449.1 | 4 - 5 | 406 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Canis familiaris Probe spans exons |