Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_850003.5 | XP_855096.1 | 2 | 386 | 172 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |