Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GJC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621072.3 | XP_005621129.1 | 217 | 181 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris |