Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197190.1 | NP_001184119.1 | 6 - 7 | 689 | 175 | |
GenBank mRNA | DN393015.1 | - | 3 - 4 | 300 | 175 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |